ENA497682770 Enamine

6-(5-chlorothiophen-2-yl)pyridazin-3-amine

  • MDL number MFCD16308285
  • Cas Number : 1225953-04-7
  • Molecular Weight : 211.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268557
Availability details for
Quantity
  • Availability
  • Add to Cart