ENA497682771 Enamine

{bicyclo[2.2.2]octan-2-yl}methanol

  • MDL number MFCD28654991
  • Cas Number : 5019-91-0
  • Molecular Weight : 140.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268005
Availability details for
Quantity
  • Availability
  • Add to Cart