ENA497682780 Enamine

(1r,4r)-4-methylcyclohexane-1-carbaldehyde

  • MDL number MFCD24675534

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-268590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268590
Availability details for
Quantity
  • Availability
  • Add to Cart