ENA497682783 Enamine

[4-(4-chlorophenoxy)phenyl]boronic acid

  • MDL number MFCD13182407
  • Cas Number : 1035491-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-268634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268634
Availability details for
Quantity
  • Availability
  • Add to Cart