ENA497682803 Enamine

4-(3-methyl-5-phenyl-1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD06742763
  • Cas Number : 93323-55-8
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-269196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269196
Availability details for
Quantity
  • Availability
  • Add to Cart