ENA497682810 Enamine

2-(3-methylbutoxy)aniline

  • MDL number MFCD06254772
  • Cas Number : 111299-83-3
  • Molecular Weight : 179.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-269212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269212
Availability details for
Quantity
  • Availability
  • Add to Cart