ENA497682811 Enamine

2-(4-methyl-1H-pyrazol-1-yl)phenol

  • MDL number MFCD20326193
  • Cas Number : 1340241-07-7
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-269214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269214
Availability details for
Quantity
  • Availability
  • Add to Cart