ENA497682814 Enamine

bicyclo[4.1.0]heptane-2-carbonitrile

  • MDL number MFCD28654138
  • Cas Number : 1781586-03-5
  • Molecular Weight : 121.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-269225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269225
Availability details for
Quantity
  • Availability
  • Add to Cart