ENA497682835 Enamine

4-methyl-1-oxo-1H-isochromene-3-carboxylic acid

  • MDL number MFCD18432365
  • Cas Number : 56661-74-6
  • Molecular Weight : 204.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-269294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-269294
Availability details for
Quantity
  • Availability
  • Add to Cart