ENA497682841 Enamine

5-bromo-2-[3-(trifluoromethyl)phenoxy]pyrimidine

  • MDL number MFCD19198907
  • Cas Number : 73226-50-3
  • Molecular Weight : 319.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268034
Availability details for
Quantity
  • Availability
  • Add to Cart