ENA497682842 Enamine

3-hydroxyphenyl N-ethylcarbamate

  • MDL number MFCD29907342
  • Cas Number : 108490-80-8
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268036
Availability details for
Quantity
  • Availability
  • Add to Cart