ENA497682846 Enamine

1-[(oxolan-2-yl)methyl]piperidin-4-one

  • MDL number MFCD09945908
  • Cas Number : 71106-10-0
  • Molecular Weight : 183.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267834
Availability details for
Quantity
  • Availability
  • Add to Cart