ENA497682857 Enamine

(3-bromo-2-methylphenyl)methanesulfonyl chloride

  • MDL number MFCD26064637
  • Cas Number : 1521597-53-4
  • Molecular Weight : 283.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268118

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-268118
pictograms GHS05,GHS07
hstatements H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart