ENA497682864 Enamine

4-chloro-2-cyclopropyl-5-methoxyaniline

  • MDL number MFCD22690118
  • Cas Number : 1383852-06-9
  • Molecular Weight : 197.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-268138
Availability details for
Quantity
  • Availability
  • Add to Cart