ENA497682930 Enamine

3,3,3-trifluoro-2-hydroxy-2-methylpropanethioamide

  • MDL number MFCD20708677
  • Cas Number : 422-25-3
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-267868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-267868
Availability details for
Quantity
  • Availability
  • Add to Cart