ENA497682932 Enamine

1-tert-butyl-4-(hydroxymethyl)pyrrolidin-2-one

  • MDL number MFCD06804425
  • Cas Number : 59857-89-5
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 64-65
Iupac Name 1-tert-butyl-4-(hydroxymethyl)-2-pyrrolidinone
Inchi Code 1S/C9H17NO2/c1-9(2,3)10-5-7(6-11)4-8(10)12/h7,11H,4-6H2,1-3H3
Inchi Key PNKQVVLPNVNTFP-UHFFFAOYSA-N

Description

Melting Point 64-65
Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-268223

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-268223
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart