ENA497682960 Enamine

4-(4-nitrophenyl)-1H-imidazol-2-amine

  • MDL number MFCD11901337
  • Cas Number : 60472-21-1
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-45134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45134
Availability details for
Quantity
  • Availability
  • Add to Cart