ENA497682999 Enamine

4-(4-chloroquinazolin-2-yl)-N,N-dimethylaniline

  • MDL number MFCD13464749
  • Cas Number : 79916-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67831
Availability details for
Quantity
  • Availability
  • Add to Cart