ENA497683021 Enamine

5-chloro-4-(difluoromethyl)-1-methyl-1H-pyrazole

  • MDL number MFCD29907468
  • Cas Number : 1909337-29-6
  • Molecular Weight : 166.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-79791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79791
Availability details for
Quantity
  • Availability
  • Add to Cart