ENA497683041 Enamine

tert-butyl 2-oxo-2,3,4,5-tetrahydro-1H-1,4-benzodiazepine-4-carboxylate

  • MDL number MFCD21222599
  • Cas Number : 179686-66-9
  • Molecular Weight : 262.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-89810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-89810
Availability details for
Quantity
  • Availability
  • Add to Cart