ENA497683075 Enamine

5-methylcyclohex-2-en-1-one

  • MDL number MFCD02685515
  • Cas Number : 7214-50-8
  • Molecular Weight : 110.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-98241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98241
Availability details for
Quantity
  • Availability
  • Add to Cart