ENA508624275 Enamine

ethyl 7-methyloctanoate

  • MDL number MFCD29991248
  • Cas Number : 29490-61-7
  • Molecular Weight : 186.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-219646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219646
Availability details for
Quantity
  • Availability
  • Add to Cart