ENA508624276 Enamine

3-(oxan-4-yl)-[1,2]oxazolo[5,4-b]pyridine-5-sulfonyl chloride

  • MDL number MFCD29991256
  • Cas Number : 1955506-71-4
  • Molecular Weight : 302.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-245661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-245661
Availability details for
Quantity
  • Availability
  • Add to Cart