ENA508624280 Enamine

tert-butyl 3-amino-2-methyl-2-phenylpropanoate

  • MDL number MFCD29991266
  • Cas Number : 1955505-46-0
  • Molecular Weight : 235.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-252183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-252183
Availability details for
Quantity
  • Availability
  • Add to Cart