ENA508624295 Enamine

2,6-dimethylpiperidine-3-carboxylic acid hydrochloride

  • MDL number MFCD29991307
  • Cas Number : 879661-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-258597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-258597
Availability details for
Quantity
  • Availability
  • Add to Cart