ENA508624309 Enamine

3-ethyl-7-phenyl-3H,4H,7H-pyrazolo[3,4-d][1,2,3]triazin-4-one

  • MDL number MFCD29991343
  • Cas Number : 1616613-79-6
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262167
Availability details for
Quantity
  • Availability
  • Add to Cart