ENA508624314 Enamine

3-bromo-8-fluoro-2-iodoquinoline

  • MDL number MFCD29991348
  • Cas Number : 1955553-57-7
  • Molecular Weight : 351.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262375
Availability details for
Quantity
  • Availability
  • Add to Cart