ENA508624316 Enamine

2-(2,2-dichlorocyclopropyl)ethan-1-ol

  • MDL number MFCD29991350
  • Cas Number : 97535-44-9
  • Molecular Weight : 155.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262683
Availability details for
Quantity
  • Availability
  • Add to Cart