ENA508624317 Enamine

2-(3-aminooxan-3-yl)acetic acid hydrochloride

  • MDL number MFCD29991351
  • Cas Number : 1955556-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-262781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262781
Availability details for
Quantity
  • Availability
  • Add to Cart