ENA508624322 Enamine

2-(3-bromo-4-methylphenyl)-4-methyl-1,3-oxazole

  • MDL number MFCD29991357
  • Cas Number : 1955520-18-9
  • Molecular Weight : 252.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262865

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262865
Availability details for
Quantity
  • Availability
  • Add to Cart