ENA508624325 Enamine

tert-butyl N-[1-(1,2,4-oxadiazol-5-yl)-2-phenylethyl]carbamate

  • MDL number MFCD29991360
  • Cas Number : 1955520-41-8
  • Molecular Weight : 289.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-262939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262939
Availability details for
Quantity
  • Availability
  • Add to Cart