ENA508624330 Enamine

4-[2-(tert-butoxy)-2-oxoethyl]oxane-4-carboxylic acid

  • MDL number MFCD29991365
  • Cas Number : 1955560-88-9
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263145
Availability details for
Quantity
  • Availability
  • Add to Cart