ENA508624332 Enamine

3-(methoxymethyl)piperidine-3-carboxylic acid hydrochloride

  • MDL number MFCD29991369
  • Cas Number : 1955554-55-8
  • Molecular Weight : 209.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-263378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-263378
Availability details for
Quantity
  • Availability
  • Add to Cart