ENA508624447 Enamine

9-fluoro-1H,2H,3H,4H,5H,6H-azepino[4,3-b]indol-1-one

  • MDL number MFCD24390476
  • Cas Number : 1009631-14-4
  • Molecular Weight : 218.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-182869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182869
Availability details for
Quantity
  • Availability
  • Add to Cart