ENA508624481 Enamine

(2-chloro-3-methoxyphenyl)methanol

  • MDL number MFCD18393574
  • Cas Number : 52516-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198614
Availability details for
Quantity
  • Availability
  • Add to Cart