ENA508624506 Enamine

tert-butyl octahydro-2H-pyrido[4,3-b]morpholine-4-carboxylate

  • MDL number MFCD28390074
  • Cas Number : 1779577-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-209746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-209746
Availability details for
Quantity
  • Availability
  • Add to Cart