ENA508624522 Enamine

2-(quinolin-4-yl)ethan-1-ol

  • MDL number MFCD11109971
  • Cas Number : 55908-35-5
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-220507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220507
Availability details for
Quantity
  • Availability
  • Add to Cart