ENA508624558 Enamine

(1-cyclopropylazetidin-2-yl)methanol

  • MDL number MFCD28654351
  • Cas Number : 1784914-12-0
  • Molecular Weight : 127.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-246430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-246430
Availability details for
Quantity
  • Availability
  • Add to Cart