ENA508624575 Enamine

3-chloro-6-(1-methyl-1H-pyrazol-5-yl)pyridazine

  • MDL number MFCD21276681
  • Cas Number : 1472021-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-250172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-250172
Availability details for
Quantity
  • Availability
  • Add to Cart