ENA508624590 Enamine

methyl 4-methyl-2-(trifluoromethyl)pentanoate

  • MDL number MFCD29991265
  • Cas Number : 1955522-87-8
  • Molecular Weight : 198.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-252151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-252151
Availability details for
Quantity
  • Availability
  • Add to Cart