ENA508624605 Enamine

tert-butyl N-[(5,6-dimethylpiperidin-2-yl)methyl]carbamate

  • MDL number MFCD29991271
  • Cas Number : 1934550-27-2
  • Molecular Weight : 242.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-253013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-253013
Availability details for
Quantity
  • Availability
  • Add to Cart