ENA508624660 Enamine

1-(4-methoxyphenyl)-5-oxopyrrolidine-3-carboxamide

  • MDL number MFCD02852918
  • Cas Number : 443638-17-3
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-255329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-255329
Availability details for
Quantity
  • Availability
  • Add to Cart