ENA508624676 Enamine

tert-butyl 2,3,5-trimethylpiperazine-1-carboxylate

  • MDL number MFCD29991280
  • Cas Number : 1936555-43-9
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-255627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-255627
Availability details for
Quantity
  • Availability
  • Add to Cart