ENA508624693 Enamine

1-(bromomethyl)cyclopentane-1-carbonitrile

  • MDL number MFCD29991289
  • Cas Number : 1936174-22-9
  • Molecular Weight : 188.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-256038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-256038
Availability details for
Quantity
  • Availability
  • Add to Cart