ENA508624719 Enamine

3-cyclopropoxy-2-fluorobenzaldehyde

  • MDL number MFCD17009447
  • Cas Number : 1243385-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-257644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-257644
Availability details for
Quantity
  • Availability
  • Add to Cart