ENA508624730 Enamine

methyl 5-(3-methylphenyl)-1H-pyrrole-2-carboxylate

  • MDL number MFCD24612277
  • Cas Number : 1000682-37-0
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-257956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-257956
Availability details for
Quantity
  • Availability
  • Add to Cart