ENA508624731 Enamine

1-fluoro-4-trifluoromethanesulfinylbenzene

  • MDL number MFCD01009706
  • Cas Number : 942-39-2
  • Molecular Weight : 212.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-257977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-257977
Availability details for
Quantity
  • Availability
  • Add to Cart