ENA508624740 Enamine

3-(2-bromoethyl)-1,2-benzothiazole

  • MDL number MFCD29991304
  • Cas Number : 1935987-70-4
  • Molecular Weight : 242.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-258398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-258398
Availability details for
Quantity
  • Availability
  • Add to Cart