ENA508624760 Enamine

3-ethoxyoxolane-3-carbohydrazide

  • MDL number MFCD29991310
  • Cas Number : 1934892-64-4
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-259701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-259701
Availability details for
Quantity
  • Availability
  • Add to Cart