ENA508624766 Enamine

3-(3-methyl-1H-pyrazol-1-yl)butanoic acid

  • MDL number MFCD14281264
  • Cas Number : 1172234-67-1
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-260060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-260060
Availability details for
Quantity
  • Availability
  • Add to Cart